Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzenesulfonic acid hydrate, 98%

Catalog No. ac415820250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415820250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415820250 Supplier Thermo Scientific Chemicals Supplier No. 415820250
Only null left
Add to Cart

CAS: 138-42-1 | C6H5NO5S | 203.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-42-1
Molecular Formula C6H5NO5S
Molecular Weight (g/mol) 203.17
MDL Number MFCD09037353
InChI Key SPXOTSHWBDUUMT-UHFFFAOYSA-N
Synonym 4-nitrobenzenesulfonic acid, benzenesulfonic acid, 4-nitro, p-nitrobenzenesulfonic acid, 4-nitrobenzenesulfonate, p-nitrophenylsulfonic acid, 4-nitrobenzenesulphonic acid, 4-nitro-benzenesulfonic acid, benzenesulfonic acid, p-nitro, ccris 3132, acmc-1bvgi
PubChem CID 8740
IUPAC Name 4-nitrobenzene-1-sulfonic acid
SMILES OS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Melting Point 105.0°C to 112.0°C
Color Beige to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula SO3HC6H4NO2
Quantity 25 g
Formula Weight 203.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzenesulfonic acid hydrate, pract., 85%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 205-329-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.