Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzenesulfonyl chloride, 97%

Catalog No. AAA1851222
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1851222 100 g
AAA1851214 25 g
AAA1851236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1851222 Supplier Thermo Scientific Chemicals Supplier No. A1851222
  • $216.50
    $184.65 / Each of 1

    Save $31.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 98-74-8 | C6H4ClNO4S | 221.61 g/mol

4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2′-bipyridine, nbs = 4-nitrobenzenesulfonate).

Solubility
Soluble in toluene, tetrahydrofuran, methylene chloride and ethyl acetate. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-74-8
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.61
MDL Number MFCD00007445
InChI Key JXRGUPLJCCDGKG-UHFFFAOYSA-N
Synonym 4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
PubChem CID 7404
SMILES [O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 76°C to 80°C
Boiling Point 143°C to 144°C (1.5 mmHg)
Quantity 100 g
UN Number UN3261
Beilstein 746543
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene,tetrahydrofuran,methylene chloride and ethyl acetate. Insoluble in water.
IUPAC Name 4-nitrobenzene-1-sulfonyl chloride
Formula Weight 221.62
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-697-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.