Learn More
Medchemexpress LLC 4-nitrobenzoic acid-d2 | 117868-95-8 | 99.0% | 169.13 | C7H3D2NO4 | 10mg
SDP
Catalog No. 5000268960
Encompass_Preferred
4-Nitrobenzoic acid-d2 is the deuterium labeled 4-Nitrobenzoic acid (HY-Y0607) 4-Nitrobenzoic acid is a nitroaromatic compound that can be used in the synthesis of other active compounds 4-Nitrobenzoic acid is also an inhibitor agent which can be used for the recognition of the Mycobacterium tuberculosis complex and differentiation from non-tuberculous mycobacteria[1]
Catalog No. 50-002-68960 Supplier Medchemexpress LLC Supplier No. HYY0607S110MG
4-Nitrobenzoic acid-d2 is the deuterium labeled 4-Nitrobenzoic acid (HY-Y0607) 4-Nitrobenzoic acid is a nitroaromatic compound that can be used in the synthesis of other active compounds 4-Nitrobenzoic acid is also an inhibitor agent which can be used for the recognition of the Mycobacterium tuberculosis complex and differentiation from non-tuberculous mycobacteria[1]