Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. N0181100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0181100G 100 g
N018125G 25 g
N0181500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0181100G Supplier TCI America Supplier No. N0181100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-11-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007373
InChI Key VOLRSQPSJGXRNJ-UHFFFAOYSA-N
Synonym 4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
PubChem CID 66011
ChEBI CHEBI:62822
IUPAC Name 1-(bromomethyl)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 99°C
Quantity 100 g
UN Number 3261
Formula Weight 216.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Bromide
RTECSNumber XS7967000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.