Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl chloroformate, 95%

Catalog No. p-3598210
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC376040050 5 g
AC376040250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376040050 Supplier Thermo Scientific Chemicals Supplier No. 376040050
Only null left
Add to Cart

CAS: 4457-32-3 | C8H6ClNO4 | 215.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4457-32-3
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00007375
InChI Key MHSGOABISYIYKP-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloroformate, 4-nitrobenzyl carbonochloridate, carbonochloridic acid, 4-nitrophenyl methyl ester, 4-nitrobenzyl chlorocarbonate, p-nitrobenzyl chloroformate, 4-nitrocarbobenzoxychloride, 4-nitrophenyl methyl chloroformate, p-nitrobenzyloxycarbonyl chloride, 4-nitrobenzyloxycarbonyl chloride, chloroformic acid p-nitrobenzyl ester
PubChem CID 78205
IUPAC Name (4-nitrophenyl)methyl carbonochloridate
SMILES C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 32°C
Flash Point 113°C
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass Bottle
Linear Formula ClCO2CH2C6H4NO2
Refractive Index 1.5520 to 1.556
Quantity 5 g
Formula Weight 215.59
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzyl chloroformate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins
EINECSNumber 224-708-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.