Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrocatechol sulfate dipotassium salt dihydrate, 97%, Thermo Scientific™

Catalog No. AAB2332501
Change view
Click to view available options
Quantity:
0.25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2332501 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2332501 Supplier Thermo Scientific Chemicals Supplier No. B2332501

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Nitrocatechol sulfate dipotassium salt dihydrate is a chromogenic sulfatase substrate. It is used as chemical, organic intermediates.

Solubility
Soluble in water (100 mg/ml).

Notes
Light sensitive. Store away from oxidizing agents, light, heat.
TRUSTED_SUSTAINABILITY

Specifications

CAS 14528-64-4
Molecular Formula C6H5K2NO7S
Quantity 0.25g
MDL Number MFCD00149509
Beilstein 3805018
Synonym disodium 5-nitro-2-oxidobenzenesulphonate
InChI Key FRSZMHGEAYKGOQ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])OS(=O)(=O)O)O.[K].[K]
IUPAC Name (2-hydroxy-5-nitrophenyl) hydrogen sulfate;potassium
Molecular Weight (g/mol) 313.363
PubChem CID 131848279
Formula Weight 347.39 (311.36 Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Nitrocatechol sulfate dipotassium salt dihydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.