Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-nitrocinnamic acid | 619-89-6 | MFCD00007381 | 99.9% | 193.16 g/mol | C9H7NO4 | 25 G
SDP

Catalog No. 5000303019
Encompass_Preferred

4-Nitrocinnamic acid is a nitro-substituted cinnamic acid used as a research reagent and synthetic building block. It is applied in organic synthesis and materials research and is supplied with analytical characterization to support laboratory use.

  • High purity for reliable experimental results.
  • Suitable as a building block in organic synthesis.
  • Soluble in common organic solvents for easy handling.
  • Characterized by CAS 619-89-6 and molecular formula C9H7NO4.
  • Supplied in controlled pack sizes for laboratory use.

Catalog No. 50-003-03019 Supplier Medchemexpress LLC Supplier No. 25GHYW015069
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Nitrocinnamic acid is a nitro-substituted cinnamic acid used as a research reagent and synthetic building block. It is applied in organic synthesis and materials research and is supplied with analytical characterization to support laboratory use.

  • High purity for reliable experimental results.
  • Suitable as a building block in organic synthesis.
  • Soluble in common organic solvents for easy handling.
  • Characterized by CAS 619-89-6 and molecular formula C9H7NO4.
  • Supplied in controlled pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.