Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrocumene 98.0+%, TCI America™
SDP

Catalog No. N063925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N063925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N063925G Supplier TCI America Supplier No. N063925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1817-47-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00039746
InChI Key JXMYUMNAEKRMIP-UHFFFAOYSA-N
Synonym 1-isopropyl-4-nitrobenzene, p-nitrocumene, 4-nitrocumene, 4-isopropylnitrobenzene, benzene, 1-1-methylethyl-4-nitro, cumene, p-nitro, p-isopropylnitrobenzene, p-nitroisopropylbenzene, 4-nitroisopropylbenzene, 2-p-nitrophenyl propane
PubChem CID 15749
IUPAC Name 1-nitro-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 2°C
Boiling Point 142°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Nitrocumene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.