Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrodiphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. N101125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N101125G 25 g
N10115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N101125G Supplier TCI America Supplier No. N101125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 620-88-2
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00007320
InChI Key JDTMUJBWSGNMGR-UHFFFAOYSA-N
Synonym 4-nitrophenyl phenyl ether, 4-nitrodiphenyl ether, p-nitrophenyl phenyl ether, benzene, 1-nitro-4-phenoxy, p-nitrodiphenyl ether, ether, 4-nitrophenyl phenyl, 4-nitrodifenylether, ether, p-nitrophenyl phenyl, 1-nitro-4-phenoxy-benzene, 4-nitrobiphenyl ether
PubChem CID 12110
IUPAC Name 1-nitro-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 61°C
Quantity 25 g
UN Number 3077
Formula Weight 215.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrodiphenyl Ether
RTECSNumber KO2680000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.