Learn More
Cayman Chemical 4-Nitrodiphenylamine, Purity: ≥98%, CAS: 836-30-6, 1 g
SDP
Catalog No. 502945458
Encompass_Preferred
An aromatic amine antioxidant; a component of both burnt gunshot residue and unburnt gunpowder
Catalog No. 50-294-5458 Supplier Cayman Chemical Supplier No. 407291 G
An aromatic amine antioxidant; a component of both burnt gunshot residue and unburnt gunpowder