Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenol, ≥99.5%, Solstice

Catalog No. 60047456 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-456 500 g Glass bottle
60-047-455 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-456 Supplier Solstice Supplier No. 73560500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 116°C
Boiling Point 279°C
Linear Formula O2NC6H4OH
UN Number UN1663
Beilstein 1281877
Solubility Information 14.8g/L at 25°C
Formula Weight 139.11g/mol
Percent Purity ≥99.5%
Packaging Glass bottle
Vapor Pressure 0.6mmHg (120°C)
Density 1.479g/cm3
Color Yellow
pH 4.4
Quantity 500 g
Physical Form Crystalline
Chemical Name or Material 4-Nitrophenol
Show More Show Less
Hazard Statement P261-P301 + P310 + P330-P302 + P352 + P312-P304 + P340 + P312
DOTInformation 6.1 / PGIII
RTECSNumber 202-811-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.