Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenol , MilliporeSigma™ Supelco™

Catalog No. 111020077 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0077 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0077 Supplier MilliporeSigma Supelco Supplier No. 48549
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Ignition Point 541°F
CAS 100-02-7
Vapor Pressure 0.6 mmHg (120°C)
Flash Point 169°C
Physical Form Neat
Linear Formula O2 NC6H4 OH
MDL Number MFCD00007331
Melting Point 110°C to 115°C
Boiling Point 279°C
Synonym p-Nitrophenol
Solubility Soluble in water
Type Organic Pollutant Standard
Quantity 5000 mg
Formula Weight 139.11
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.