Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenoxyacetic Acid 98.0+%, TCI America™
SDP

Catalog No. N0226500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0226500G 500 g
N022625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0226500G Supplier TCI America Supplier No. N0226500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1798-11-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00017030
InChI Key AVDLFIONKHGQAP-UHFFFAOYSA-N
Synonym 4-nitrophenoxyacetic acid, 2-4-nitrophenoxy acetic acid, 4-nitrophenoxy acetic acid, p-nitrophenoxyacetic acid, acetic acid, 4-nitrophenoxy, 4-nitro-phenoxy-acetic acid, acetic acid, p-nitrophenoxy, acetic acid, 2-4-nitrophenoxy, acmc-209efx
PubChem CID 15720
SMILES C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)O

Specifications

Melting Point 187°C
Quantity 500 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenoxyacetic Acid
EINECSNumber (3)-0781
RTECSNumber AJ1100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.