Learn More
Research Products International Corp P-NITROPHENYL-A-D-GLUCOPYRANOSIDE, 1 GRAM
SDP
Catalog No. 50488878
Encompass_Preferred
Spectrophotometer substrate for Glucansucrases.
Free PNP: <250ppm
Catalog No. 50-488-878 Supplier Research Products International Corp Supplier No. N820801.0
Spectrophotometer substrate for Glucansucrases.
Free PNP: <250ppm