Learn More
Research Products International Corp 4-Nitrophenyl-N-Acetyl Glucosaminide, 1 gram
SDP
Catalog No. 50213566
Encompass_Preferred
Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.
Catalog No. 50-213-566 Supplier Research Products International Corp Supplier No. N516001.0
Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.