Learn More
Bioworld 4-Nitrophenyl-2-acetamido-2-deoxy-B-D-Galactopyranoside, 100 mg
Supplier:  Bioworld 215300241
Encompass_Preferred
C14H18N2O8 MW 342.3 Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase. * Purity: >99% * Soluble in DMSO and DMFO * Cas No. 14948-96-0 This product is sold for laboratory research use only
Catalog No. 50-198-804