Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-nitrophenyl 2-(pyridin-2-yldisulfanyl)ethyl carbonate | 874302-76-8 | MFCD28556879 | 99.6% | 352.39 g/mol | C14H12N2O5S2 | 5 G
SDP

Catalog No. 5000202956
Encompass_Preferred

PDEC-NB is a disulfide-cleavable linker used for the synthesis of antibody-drug conjugates (ADCs). It releases payloads under reducing intracellular conditions via a pyridinyl-disulfide-triggered mechanism. Supplied as a white to off-white solid for research use only.

  • Used as a disulfide cleavable linker for ADC synthesis.
  • Molecular weight 352.39 g/mol.
  • Chemical formula C14H12N2O5S2.
  • CAS number 874302-76-8.
  • Appearance white to off-white solid.
  • For research use only; not for human use.

Catalog No. 50-002-02956 Supplier Medchemexpress LLC Supplier No. HY1264985G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PDEC-NB is a disulfide-cleavable linker used for the synthesis of antibody-drug conjugates (ADCs). It releases payloads under reducing intracellular conditions via a pyridinyl-disulfide-triggered mechanism. Supplied as a white to off-white solid for research use only.

  • Used as a disulfide cleavable linker for ADC synthesis.
  • Molecular weight 352.39 g/mol.
  • Chemical formula C14H12N2O5S2.
  • CAS number 874302-76-8.
  • Appearance white to off-white solid.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.