Learn More
Medchemexpress LLC (4-nitrophenyl) 2-(pyridin-2-yldisulfanyl)ethyl carbonate | 874302-76-8 | MFCD28556879 | 99.9% | 352.4 g/mol | C14H12N2O5S2 | 250 MG
SDP
Catalog No. 5000203742
Encompass_Preferred
PDEC-NB is a disulfide-cleavable carbonate linker used to construct antibody-drug conjugates (ADCs). It contains an activated 4-nitrophenyl carbonate and a pyridin-2-yldisulfanyl moiety that enables release of payloads under reducing intracellular conditions.
  • Provides disulfide cleavable linkage for intracellular payload release
  • Contains an activated 4-nitrophenyl carbonate for efficient conjugation
  • White to off-white solid suitable for low-temperature storage
  • Available in mg to gram scale quantities for research use
  • High purity appropriate for research applications
Catalog No. 50-002-03742 Supplier Medchemexpress LLC Supplier No. HY126498250MG
PDEC-NB is a disulfide-cleavable carbonate linker used to construct antibody-drug conjugates (ADCs). It contains an activated 4-nitrophenyl carbonate and a pyridin-2-yldisulfanyl moiety that enables release of payloads under reducing intracellular conditions.
  • Provides disulfide cleavable linkage for intracellular payload release
  • Contains an activated 4-nitrophenyl carbonate for efficient conjugation
  • White to off-white solid suitable for low-temperature storage
  • Available in mg to gram scale quantities for research use
  • High purity appropriate for research applications