Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Nitrophenyl 2-(Trimethylsilyl)Ethyl Carbonate

Catalog No. 927485G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-748-5G 5 g
92-748-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 92-748-5G Supplier Sigma Aldrich Supplier No. 927485G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Nitrophenyl 2-(Trimethylsilyl)Ethyl Carbonate

Specifications

CAS 80149-80-0
Melting Point 35°C to 40°C
Molecular Formula C12H17NO5Si
Linear Formula C12H17NO5Si
Quantity 5 g
MDL Number MFCD00042930
Synonym 2-(Trimethylsilyl)ethyl 4-nitrophenyl carbonate; TEOC-ONp
Molecular Weight (g/mol) 283.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.