Learn More
Research Products International Corp 4-Nitrophenyl-N-Acetyl Glucosaminide 250mg
SDP
Catalog No. 50213565
Encompass_Preferred
Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.
Catalog No. 50-213-565 Supplier Research Products International Corp Supplier No. N516000.25
Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.