Learn More
Medchemexpress LLC 4-Nitrophenyl 4-guanidinobenzoate hydrochloride | 19135-17-2 | 99.38% | 336.73 | 25 MG
SDP
Catalog No. 5000351917
Encompass_Preferred
4-Nitrophenyl 4-guanidinobenzoate (NPGB) hydrochloride is a potent sperm acrosin inhibitor, and also a trypsin substrate.
  • Potent sperm acrosin inhibitor.
  • Trypsin substrate.
Catalog No. 50-003-51917 Supplier Medchemexpress LLC Supplier No. HY13781450MG
4-Nitrophenyl 4-guanidinobenzoate (NPGB) hydrochloride is a potent sperm acrosin inhibitor, and also a trypsin substrate.
  • Potent sperm acrosin inhibitor.
  • Trypsin substrate.