Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Nitrophenyl 4-guanidinobenzoate hydrochloride | 19135-17-2 | 99.4% | 336.73 | 100 MG
SDP

Catalog No. 5000351442
Encompass_Preferred

4-Nitrophenyl 4-guanidinobenzoate hydrochloride (NPGB hydrochloride) is a potent sperm acrosin inhibitor and also a trypsin substrate. This chemical is supplied as a solid, typically white to off-white in color, with a high purity of 99.38%.

  • Potent sperm acrosin inhibitor
  • Also functions as a trypsin substrate
  • Appearance as a solid
  • Color ranges from white to off-white
  • High purity of 99.38%
  • Molecular weight of 336.73
  • Store at 4°C, sealed, away from moisture

Catalog No. 50-003-51442 Supplier Medchemexpress LLC Supplier No. HY137814100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Nitrophenyl 4-guanidinobenzoate hydrochloride (NPGB hydrochloride) is a potent sperm acrosin inhibitor and also a trypsin substrate. This chemical is supplied as a solid, typically white to off-white in color, with a high purity of 99.38%.

  • Potent sperm acrosin inhibitor
  • Also functions as a trypsin substrate
  • Appearance as a solid
  • Color ranges from white to off-white
  • High purity of 99.38%
  • Molecular weight of 336.73
  • Store at 4°C, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.