Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Nitrophenyl a-D-glucopyranoside | 3767-28-0 | 99.8% | 301.25 | 10MM/1ML
SDP

Catalog No. 5000454907
Encompass_Preferred

4-Nitrophenyl a-D-glucopyranoside is a chromogenic substrate for α-glucosidase and can be used to measure α-glucosidase activity. It releases p-nitrophenol by enzymatic cleavage, which can be quantified by colorimetric detection at 405 nm.

  • Purity: 99.78%
  • CAS number: 3767-28-0
  • Molecular weight: 301.25
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 50 mg/mL
  • Solubility in H2O: 11.67 mg/mL

Catalog No. 50-004-54907 Supplier Medchemexpress LLC Supplier No. HYW01141110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Nitrophenyl a-D-glucopyranoside is a chromogenic substrate for α-glucosidase and can be used to measure α-glucosidase activity. It releases p-nitrophenol by enzymatic cleavage, which can be quantified by colorimetric detection at 405 nm.

  • Purity: 99.78%
  • CAS number: 3767-28-0
  • Molecular weight: 301.25
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 50 mg/mL
  • Solubility in H2O: 11.67 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.