Learn More
Research Products International Corp 4-NITROPHENYL-A-D-MANNOPYRANOSIDE 500MG
SDP
Catalog No. 50213579
Encompass_Preferred
Spectrophotometric substrate for detection of α-Mannosidase.
Free PNP: <200ppm
Catalog No. 50-213-579 Supplier Research Products International Corp Supplier No. N810600.5
Spectrophotometric substrate for detection of α-Mannosidase.
Free PNP: <200ppm