Learn More
Research Products International Corp P-NITROPHENYL-A-D-MANNOPYRANOSIDE, 1 GRAM
SDP
Catalog No. 50213580
Encompass_Preferred
Spectrophotometric substrate for detection of α-Mannosidase.
Free PNP: <200ppm
Catalog No. 50-213-580 Supplier Research Products International Corp Supplier No. N810601.0
Spectrophotometric substrate for detection of α-Mannosidase.
Free PNP: <200ppm