Learn More
Medchemexpress LLC 4-nitrophenyl alpha-D-galactopyranoside | 7493-95-0 | MFCD00065050 | >98.0% | 301.25 g/mol | C12H15NO8 | 250 MG
SDP
Catalog No. 5000303062
Encompass_Preferred
4-Nitrophenyl α-D-galactopyranoside is a chromogenic substrate used to measure α-galactosidase activity. Enzymatic hydrolysis liberates p-nitrophenol, producing a yellow product that is quantified spectrophotometrically (≈405 nm), enabling straightforward colorimetric and kinetic assays in biochemical research and enzymology.
Catalog No. 50-003-03062 Supplier Medchemexpress LLC Supplier No. 250MGHYW039911
4-Nitrophenyl α-D-galactopyranoside is a chromogenic substrate used to measure α-galactosidase activity. Enzymatic hydrolysis liberates p-nitrophenol, producing a yellow product that is quantified spectrophotometrically (≈405 nm), enabling straightforward colorimetric and kinetic assays in biochemical research and enzymology.