Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl alpha-D-Galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. N04921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04921G 1 g
N0492200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04921G Supplier TCI America Supplier No. N04921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7493-95-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00065050
InChI Key IFBHRQDFSNCLOZ-IIRVCBMXSA-N
Synonym 4-nitrophenyl a-d-galactopyranoside, 4-nitrophenyl-alpha-d-galactopyranoside, pnpalphagal, p-nitrophenyl-alpha-d-galactopyranoside, 4-nitrophenyl, a-d-galactopyranoside, p-nitrophenyl alpha-d-galactopyranoside, 4-nitrophenyl alpha-d-galactopyranoside, p-nitrophenyl-alpha-d-galactoside, 4-nitrophenyl-alpha-d-galactoside, 1-o-4-nitrophenyl-alpha-d-galactose
PubChem CID 82000
ChEBI CHEBI:546840
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 1 g
Formula Weight 301.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl alpha-D-Galactopyranoside [Substrate for alpha-D-Galactosidase]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.