Learn More
Medchemexpress LLC 4-nitrophenyl alpha-D-glucopyranoside | 3767-28-0 | 99.8% | 301.25 | 100 MG
SDP
Catalog No. 5000454926
Encompass_Preferred
4-Nitrophenyl α-D-glucopyranoside is a chromogenic substrate used for α-glucosidase, allowing for the measurement of α-glucosidase activity. It releases p-nitrophenol through enzymatic cleavage, which can be quantified colorimetrically at 405 nm.
  • It is a chromogenic substrate for α-glucosidase.
  • It can be used to measure α-glucosidase activity.
  • It releases p-nitrophenol by enzymatic cleavage.
  • P-nitrophenol can be quantified by colorimetric detection at 405 nm.
Catalog No. 50-004-54926 Supplier Medchemexpress LLC Supplier No. HYW011411100MG
4-Nitrophenyl α-D-glucopyranoside is a chromogenic substrate used for α-glucosidase, allowing for the measurement of α-glucosidase activity. It releases p-nitrophenol through enzymatic cleavage, which can be quantified colorimetrically at 405 nm.
  • It is a chromogenic substrate for α-glucosidase.
  • It can be used to measure α-glucosidase activity.
  • It releases p-nitrophenol by enzymatic cleavage.
  • P-nitrophenol can be quantified by colorimetric detection at 405 nm.