Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl alpha-D-Glucuronide 97.0+%, TCI America™
SDP

Catalog No. N085725MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N085725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N085725MG Supplier TCI America Supplier No. N085725MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71484-85-0
Molecular Formula C12H13NO9
Molecular Weight (g/mol) 315.234
InChI Key QSUILVWOWLUOEU-GBVMLCMLSA-N
Synonym 4-Nitrophenyl alpha-D-Glucopyranosiduronic Acid
PubChem CID 53384364
IUPAC Name (2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylic acid
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)C(=O)O)O)O)O

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 315.23
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl alpha-D-Glucuronide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.