Learn More
Research Products International Corp 4-Nitrophenyl-B-D-Glucopyranoside, 1 Gram
SDP
Catalog No. 50213581
Encompass_Preferred
Spectrophotometric substrate for detection of β-D-Glucosidase.
Free PNP: <200ppm
Catalog No. 50-213-581 Supplier Research Products International Corp Supplier No. N810901.0
Spectrophotometric substrate for detection of β-D-Glucosidase.
Free PNP: <200ppm