Learn More
Research Products International Corp 4-Nitrophenyl-B-D-Glucopyranoside, 5 Grams
SDP
Catalog No. 50213582
Encompass_Preferred
Spectrophotometric substrate for detection of β-D-Glucosidase.
Free PNP: <200ppm
Catalog No. 50-213-582 Supplier Research Products International Corp Supplier No. N810905.0
Spectrophotometric substrate for detection of β-D-Glucosidase.
Free PNP: <200ppm