Learn More
Cayman Chemical 4-Nitrophenyl Butyrate, Purity: ≥95%, CAS: 2635-84-9, 10 g
SDP
Catalog No. 502936082
Encompass_Preferred
A colorimetric substrate for lipases; converted into p-nitrophenoxide, which can be monitored using UV-Vis spectrophotometry at 400 nm to quantify lipase activity
Catalog No. 50-293-6082 Supplier Cayman Chemical Supplier No. 2543910 G
A colorimetric substrate for lipases; converted into p-nitrophenoxide, which can be monitored using UV-Vis spectrophotometry at 400 nm to quantify lipase activity