Learn More
Medchemexpress LLC 4-nitrophenyl butyrate | 2635-84-9 | MFCD00047734 | 99.9% | 209.20 g/mol | C10H11NO4 | 1 G
SDP
Catalog No. 5000302912
Encompass_Preferred
4-Nitrophenyl butyrate is a chromogenic ester substrate commonly used to measure esterase and lipase activity in biochemical assays. Enzymatic hydrolysis releases 4-nitrophenol, producing a color change that can be monitored spectrophotometrically for kinetic or end-point measurements. It is also used as an organic synthesis reagent and as an intermediate in dye chemistry. Supplied for research use only.
  • Chromogenic substrate for esterase and lipase assays
  • Enzymatic hydrolysis yields 4-nitrophenol for colorimetric detection
  • High purity suitable for analytical applications
  • Stable liquid form with defined density and appearance
  • Applicable as a reagent in organic synthesis and dye intermediates
Catalog No. 50-003-02912 Supplier Medchemexpress LLC Supplier No. HYW0144491G
4-Nitrophenyl butyrate is a chromogenic ester substrate commonly used to measure esterase and lipase activity in biochemical assays. Enzymatic hydrolysis releases 4-nitrophenol, producing a color change that can be monitored spectrophotometrically for kinetic or end-point measurements. It is also used as an organic synthesis reagent and as an intermediate in dye chemistry. Supplied for research use only.
  • Chromogenic substrate for esterase and lipase assays
  • Enzymatic hydrolysis yields 4-nitrophenol for colorimetric detection
  • High purity suitable for analytical applications
  • Stable liquid form with defined density and appearance
  • Applicable as a reagent in organic synthesis and dye intermediates