Learn More
Medchemexpress LLC 4-Nitrophenyl β-D-cellobioside | 3482-57-3 | 99.9% | 463.39 | 100 MG
SDP
Catalog No. 5000350587
Encompass_Preferred
4-Nitrophenyl β-D-Cellobioside, also known as p-Nitrophenyl β-D-cellobioside, is a cellotriose analog. It can be hydrolyzed by β-glucosidases like TxGH116 and ThCel7B, as well as by exoglucanases and endoglucanases, to produce p-nitrophenol (PNP). This compound is utilized to detect cellulase activity.
  • Cellotriose analog
  • Hydrolyzed by β-glucosidases (TxGH116, ThCel7B)
  • Hydrolyzed by exoglucanases and endoglucanases to produce p-nitrophenol (PNP)
  • Can be used to detect cellulase activity
  • High purity of 99.94%
Catalog No. 50-003-50587 Supplier Medchemexpress LLC Supplier No. HY137827100MG
4-Nitrophenyl β-D-Cellobioside, also known as p-Nitrophenyl β-D-cellobioside, is a cellotriose analog. It can be hydrolyzed by β-glucosidases like TxGH116 and ThCel7B, as well as by exoglucanases and endoglucanases, to produce p-nitrophenol (PNP). This compound is utilized to detect cellulase activity.
  • Cellotriose analog
  • Hydrolyzed by β-glucosidases (TxGH116, ThCel7B)
  • Hydrolyzed by exoglucanases and endoglucanases to produce p-nitrophenol (PNP)
  • Can be used to detect cellulase activity
  • High purity of 99.94%