Learn More
Medchemexpress LLC 4-Nitrophenyl β-D-Cellobioside | 3482-57-3 | 99.9% | 463.39 | 50 MG
SDP
Catalog No. 5000350685
Encompass_Preferred
4-Nitrophenyl β-D-Cellobioside is a cellotriose analog used to detect cellulase activity. It is hydrolyzed by β-glucosidases and other glycosidases to produce p-nitrophenol (PNP), making it suitable for various enzymatic assays.
  • Cellotriose analog
  • Hydrolyzed by β-glucosidases
  • Hydrolyzed by exoglucanases and endoglucanases
  • Produces p-nitrophenol (PNP)
  • Detects cellulase activity
Catalog No. 50-003-50685 Supplier Medchemexpress LLC Supplier No. HY13782750MG
4-Nitrophenyl β-D-Cellobioside is a cellotriose analog used to detect cellulase activity. It is hydrolyzed by β-glucosidases and other glycosidases to produce p-nitrophenol (PNP), making it suitable for various enzymatic assays.
  • Cellotriose analog
  • Hydrolyzed by β-glucosidases
  • Hydrolyzed by exoglucanases and endoglucanases
  • Produces p-nitrophenol (PNP)
  • Detects cellulase activity