Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl-β-D-fucopyranoside, Thermo Scientific™

Catalog No. AC336851000
Change view
Click to view available options
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC336851000 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC336851000 Supplier Thermo Scientific Chemicals Supplier No. 336851000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1226-39-7
Packaging Glass bottle
Molecular Formula C12H15NO7
Quantity 100mg
MDL Number MFCD00040651
Synonym 4-nitrophenyl beta-d-fucopyranoside, 4-nitrophenyl b-d-fucopyranoside, 4-nitrophenyl-beta-d-fucopyranoside, p-nitrophenyl beta-d-fucopyranoside, 2r,3r,4s,5r,6s-2-methyl-6-4-nitrophenoxy oxane-3,4,5-triol, b-d-galactopyranoside,4-nitrophenyl 6-deoxy, 4-nitrophenyl 6-deoxy-beta-d-galactopyranoside, 4-nitrophenyl, a-d-fucopyranoside, 2-nitrophenyl i 2-d-fucopyranoside, 4-nitrophenyl i 2-d-fucopyranoside
InChI Key YILIDCGSXCGACV-BVWHHUJWSA-N
SMILES CC1C(C(C(C(O1)OC2=CC=C(C=C2)[N+](=O)[O-])O)O)O
IUPAC Name (2R,3R,4S,5R,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol) 285.252
PubChem CID 2733769
Formula Weight 285.25
Chemical Name or Material 4-Nitrophenyl-β-D-fucopyranoside
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.