Learn More
Bioworld 4-Nitrophenyl-B-D-Galactopyranoside, 1 g
Catalog No. 50198809
Encompass_Preferred
Colorimetric and spectrophotoametric substrate for detection of β-D Galactosidase. * Purity: >99% * Soluble in water * Protect from light and moisture * Cas No. 3150-24-1 This product is sold for laboratory research use only.
Catalog No. 50-198-809 Supplier Bioworld Supplier No. 215300261
Colorimetric and spectrophotoametric substrate for detection of β-D Galactosidase. * Purity: >99% * Soluble in water * Protect from light and moisture * Cas No. 3150-24-1 This product is sold for laboratory research use only.