Learn More
Medchemexpress LLC 4-nitrophenyl α-D-galactopyranoside | 7493-95-0 | MFCD00065050 | 98.7% | 301.25 g/mol | C12H15NO8 | 100 MG
SDP
Catalog No. 5000301250
Encompass_Preferred
4-Nitrophenyl α-D-galactopyranoside is a chromogenic artificial substrate used to detect and quantify α-galactosidase activity. Cleavage by α-galactosidase releases p-nitrophenol, producing a colorimetric signal suitable for enzyme assays and activity screening in biochemical and glycobiology research.
  • Releases p-nitrophenol (pNP) upon enzymatic cleavage for colorimetric detection.
  • Suitable for quantitative α-galactosidase activity assays.
  • High purity supports reproducible assay results.
  • Available in small quantity sizes for research use.
  • Powder form with recommended cold storage to maintain stability.
Catalog No. 50-003-01250 Supplier Medchemexpress LLC Supplier No. HYW039911100MG
4-Nitrophenyl α-D-galactopyranoside is a chromogenic artificial substrate used to detect and quantify α-galactosidase activity. Cleavage by α-galactosidase releases p-nitrophenol, producing a colorimetric signal suitable for enzyme assays and activity screening in biochemical and glycobiology research.
  • Releases p-nitrophenol (pNP) upon enzymatic cleavage for colorimetric detection.
  • Suitable for quantitative α-galactosidase activity assays.
  • High purity supports reproducible assay results.
  • Available in small quantity sizes for research use.
  • Powder form with recommended cold storage to maintain stability.