Learn More
Medchemexpress LLC 4-Nitrophenyl α-D-glucopyranoside | 3767-28-0 | 99.8% | 301.25 | 500 MG
SDP
Catalog No. 5000455072
Encompass_Preferred
4-Nitrophenyl α-D-glucopyranoside is a chromogenic substrate designed for α-glucosidase. It can be utilized to measure α-glucosidase activity.
  • Chromogenic substrate for α-glucosidase.
  • Measures α-glucosidase activity.
  • Releases p-nitrophenol by enzymatic cleavage.
  • Quantifiable by colorimetric detection at 405 nm.
  • For research use only.
Catalog No. 50-004-55072 Supplier Medchemexpress LLC Supplier No. HYW011411500MG
4-Nitrophenyl α-D-glucopyranoside is a chromogenic substrate designed for α-glucosidase. It can be utilized to measure α-glucosidase activity.
  • Chromogenic substrate for α-glucosidase.
  • Measures α-glucosidase activity.
  • Releases p-nitrophenol by enzymatic cleavage.
  • Quantifiable by colorimetric detection at 405 nm.
  • For research use only.