Learn More
Alkali Scientific 4 Nitrophenyl β D Glucopyranoside, 5 G
SDP
Catalog No. 5000661815
Encompass_Preferred
4-Nitrophenyl β-D-Glucopyranoside is a chromogenic substrate used to detect and measure β-glucosidase activity. The 5g pack is suitable for a range of laboratory applications, from small-scale experiments to routine enzyme assays. This substrate undergoes a color change when hydrolyzed by β-glucosidase, providing a simple, visual method for detecting enzymatic activity. It is widely used in studies of carbohydrate metabolism and enzymatic function, particularly in microbiology and biochemistry. 4-Nitrophenyl β-D-Glucopyranoside is a valuable tool in enzyme kinetics and is also used in diagnostic tests to assess β-glucosidase activity in biological samples.
Catalog No. 50-006-61815 Supplier Alkali Scientific Supplier No. TRO002CP
4-Nitrophenyl β-D-Glucopyranoside is a chromogenic substrate used to detect and measure β-glucosidase activity. The 5g pack is suitable for a range of laboratory applications, from small-scale experiments to routine enzyme assays. This substrate undergoes a color change when hydrolyzed by β-glucosidase, providing a simple, visual method for detecting enzymatic activity. It is widely used in studies of carbohydrate metabolism and enzymatic function, particularly in microbiology and biochemistry. 4-Nitrophenyl β-D-Glucopyranoside is a valuable tool in enzyme kinetics and is also used in diagnostic tests to assess β-glucosidase activity in biological samples.