Learn More
Cayman Chemical 4-Nitrophenyl β-D-Glucopyranoside, Purity: ≥98%, CAS: 2492-87-7, 1 g
SDP
Catalog No. 502911439
Encompass_Preferred
A chromogenic β-glucosidase substrate; upon hydrolysis by β-glucosidase, 4-nitrophenol is released; can be quantified by colorimetric detection at 405 nm as a measure of β-glucosidase activity
Catalog No. 50-291-1439 Supplier Cayman Chemical Supplier No. 351091 G
A chromogenic β-glucosidase substrate; upon hydrolysis by β-glucosidase, 4-nitrophenol is released; can be quantified by colorimetric detection at 405 nm as a measure of β-glucosidase activity