Learn More
Medchemexpress LLC 4-nitrophenyl β-D-glucuronide | 10344-94-2 | MFCD00036210 | 99.1% | 315.23 g/mol | C12H13NO9 | 1 ML
SDP
Catalog No. 5000299826
Encompass_Preferred
4-Nitrophenyl β-D-glucuronide is a chromogenic substrate used to measure β-glucuronidase activity. Enzymatic hydrolysis releases 4-nitrophenol, producing a quantifiable yellow product for spectrophotometric assays. Supplied as a solid or as a 10 mM solution in DMSO for enzymology and biochemical applications.
  • Chromogenic substrate for β-glucuronidase assays.
  • Releases 4-nitrophenol for spectrophotometric detection.
  • Available as solid and as 10 mM solution in DMSO.
  • High purity suitable for enzymatic assays.
  • Documentation available: data sheet, COA, SDS, and spectral data.
Catalog No. 50-002-99826 Supplier Medchemexpress LLC Supplier No. HYW03989210MM/1ML
4-Nitrophenyl β-D-glucuronide is a chromogenic substrate used to measure β-glucuronidase activity. Enzymatic hydrolysis releases 4-nitrophenol, producing a quantifiable yellow product for spectrophotometric assays. Supplied as a solid or as a 10 mM solution in DMSO for enzymology and biochemical applications.
  • Chromogenic substrate for β-glucuronidase assays.
  • Releases 4-nitrophenol for spectrophotometric detection.
  • Available as solid and as 10 mM solution in DMSO.
  • High purity suitable for enzymatic assays.
  • Documentation available: data sheet, COA, SDS, and spectral data.