Learn More
Alkali Scientific 4 Nitrophenyl β D xylopyranoside, 1 G
SDP
Catalog No. 5000659901
Encompass_Preferred
4-Nitrophenyl β-D-xylopyranoside, 1g, is a widely used reagent in the biochemical analysis of xylanase and glycosidase activities. When hydrolyzed by these enzymes, it releases a p-nitrophenol group that can be detected using spectrophotometric methods. This compound is essential in studies focused on the breakdown of hemicellulose, particularly in biofuel production, where xylanases play a significant role in converting plant biomass into fermentable sugars. The 1g quantity is suitable for medium-scale experiments and is used in routine enzyme assays, carbohydrate metabolism studies, and biotechnological applications.
Catalog No. 50-006-59901 Supplier Alkali Scientific Supplier No. LBE002CP
4-Nitrophenyl β-D-xylopyranoside, 1g, is a widely used reagent in the biochemical analysis of xylanase and glycosidase activities. When hydrolyzed by these enzymes, it releases a p-nitrophenol group that can be detected using spectrophotometric methods. This compound is essential in studies focused on the breakdown of hemicellulose, particularly in biofuel production, where xylanases play a significant role in converting plant biomass into fermentable sugars. The 1g quantity is suitable for medium-scale experiments and is used in routine enzyme assays, carbohydrate metabolism studies, and biotechnological applications.