Learn More
Medchemexpress LLC 4-nitrophenyl β-D-xylopyranoside | 2001-96-9 | MFCD00047519 | 99.9% | 271.22 g/mol | C11H13NO7 | 100 MG
SDP
Catalog No. 5000299494
Encompass_Preferred
4-Nitrophenyl β-D-xylopyranoside is a chromogenic β-xylosidase substrate that releases p-nitrophenol upon enzymatic cleavage, enabling sensitive colorimetric measurement of β-xylosidase activity. Supplied as a solid (100 mg), it is suitable for biochemical assays and enzyme characterization.
Catalog No. 50-002-99494 Supplier Medchemexpress LLC Supplier No. HYW039938100MG
4-Nitrophenyl β-D-xylopyranoside is a chromogenic β-xylosidase substrate that releases p-nitrophenol upon enzymatic cleavage, enabling sensitive colorimetric measurement of β-xylosidase activity. Supplied as a solid (100 mg), it is suitable for biochemical assays and enzyme characterization.