Learn More
Cayman Chemical 4-Nitrophenyl β-D-Xylopyranoside, Purity: ≥98%, CAS: 2001-96-9, 1 g
SDP
Catalog No. 502919070
Encompass_Preferred
A chromogenic β-xylosidase substrate; upon hydrolysis by β-xylosidase, 4-nitrophenol is released, which can be quantified by colorimetric detection at 405 nm as a measure of β-xylosidase activity
Catalog No. 50-291-9070 Supplier Cayman Chemical Supplier No. 388831 G
A chromogenic β-xylosidase substrate; upon hydrolysis by β-xylosidase, 4-nitrophenol is released, which can be quantified by colorimetric detection at 405 nm as a measure of β-xylosidase activity