Learn More
Alkali Scientific 4 Nitrophenyl α L Arabinopyranoside, 500 Mg
SDP
Catalog No. 5000661890
Encompass_Preferred
4 Nitrophenyl α-L Arabinopyranoside (500 mg) is a potent substrate used to study the activity of α-L-arabinosidases in biochemical assays. The compound, upon hydrolysis by the enzyme, releases a yellow color, providing an easy-to-detect indicator of enzyme activity. It is widely used in glycobiology and carbohydrate chemistry to study the breakdown of arabinose-containing polysaccharides, such as hemicelluloses. This reagent is commonly applied in enzyme research, high-throughput screening, and the development of novel α-L-arabinosidase inhibitors. It is also valuable in exploring the role of arabinose in various cellular processes, including metabolism and signaling.
Catalog No. 50-006-61890 Supplier Alkali Scientific Supplier No. RAB002CP
4 Nitrophenyl α-L Arabinopyranoside (500 mg) is a potent substrate used to study the activity of α-L-arabinosidases in biochemical assays. The compound, upon hydrolysis by the enzyme, releases a yellow color, providing an easy-to-detect indicator of enzyme activity. It is widely used in glycobiology and carbohydrate chemistry to study the breakdown of arabinose-containing polysaccharides, such as hemicelluloses. This reagent is commonly applied in enzyme research, high-throughput screening, and the development of novel α-L-arabinosidase inhibitors. It is also valuable in exploring the role of arabinose in various cellular processes, including metabolism and signaling.