Learn More
Medchemexpress LLC 4-Nitrophenyl α-L-rhamnopyranoside | 18918-31-5 | 99.8% | 285.25 | 1 ML
SDP
Catalog No. 5000352190
Encompass_Preferred
4-Nitrophenyl α-L-rhamnopyranoside is a commonly used substrate in various biochemical assays to measure the activity of enzymes that hydrolyze rhamnose, such as α-L-rhamnosidase. It has unique chemical properties that allow it to be hydrolyzed by these enzymes to form a yellow product called p-nitrophenol, making it a useful tool for detecting and quantifying rhamnohydrolase activity in biological samples or microbial cultures.
  • Commonly used substrate in biochemical assays
  • Measures activity of enzymes that hydrolyze rhamnose
  • Hydrolyzed to form a yellow product, p-nitrophenol
  • Useful for detecting and quantifying rhamnohydrolase activity
  • Applicable in biological samples or microbial cultures
Catalog No. 50-003-52190 Supplier Medchemexpress LLC Supplier No. HY13781510MM/1ML
4-Nitrophenyl α-L-rhamnopyranoside is a commonly used substrate in various biochemical assays to measure the activity of enzymes that hydrolyze rhamnose, such as α-L-rhamnosidase. It has unique chemical properties that allow it to be hydrolyzed by these enzymes to form a yellow product called p-nitrophenol, making it a useful tool for detecting and quantifying rhamnohydrolase activity in biological samples or microbial cultures.
  • Commonly used substrate in biochemical assays
  • Measures activity of enzymes that hydrolyze rhamnose
  • Hydrolyzed to form a yellow product, p-nitrophenol
  • Useful for detecting and quantifying rhamnohydrolase activity
  • Applicable in biological samples or microbial cultures