Learn More
Medchemexpress LLC 4-Nitrophenyl α-L-rhamnopyranoside | 18918-31-5 | 99.8% | 285.25 | 50 MG
SDP
Catalog No. 5000351443
Encompass_Preferred
4-Nitrophenyl α-L-rhamnopyranoside is a substrate commonly used in various biochemical assays to measure the activity of enzymes that hydrolyze rhamnose, such as α-L-rhamnosidase. It possesses unique chemical properties that enable its hydrolysis by these enzymes, resulting in the formation of a yellow product called p-nitrophenol. This characteristic makes it a valuable tool for detecting and quantifying rhamnohydrolase activity in biological samples or microbial cultures.
  • Commonly used substrate in biochemical assays
  • Measures activity of enzymes that hydrolyze rhamnose, such as α-L-rhamnosidase
  • Hydrolysis forms a yellow product, p-nitrophenol
  • Valuable tool for detecting and quantifying rhamnohydrolase activity in biological samples or microbial cultures
Catalog No. 50-003-51443 Supplier Medchemexpress LLC Supplier No. HY13781550MG
4-Nitrophenyl α-L-rhamnopyranoside is a substrate commonly used in various biochemical assays to measure the activity of enzymes that hydrolyze rhamnose, such as α-L-rhamnosidase. It possesses unique chemical properties that enable its hydrolysis by these enzymes, resulting in the formation of a yellow product called p-nitrophenol. This characteristic makes it a valuable tool for detecting and quantifying rhamnohydrolase activity in biological samples or microbial cultures.
  • Commonly used substrate in biochemical assays
  • Measures activity of enzymes that hydrolyze rhamnose, such as α-L-rhamnosidase
  • Hydrolysis forms a yellow product, p-nitrophenol
  • Valuable tool for detecting and quantifying rhamnohydrolase activity in biological samples or microbial cultures