Learn More
Medchemexpress LLC 4-nitrophenyl α-L-rhamnoside | 18918-31-5 | MFCD00069788 | 99.8% | C12H15NO7 | 10MG
SDP
Catalog No. 5000261771
Encompass_Preferred
4-Nitrophenyl α-L-rhamnopyranoside is a chromogenic substrate used to detect and quantify α-L-rhamnosidase activity. Enzymatic hydrolysis releases p-nitrophenol, producing a yellow product that is convenient for spectrophotometric measurement. Supplied in high purity as a powder, it is also available as a 10 mM solution in DMSO for ready use. For research use only.
  • Chromogenic substrate for α-L-rhamnosidase assays.
  • Releases p-nitrophenol on hydrolysis for colorimetric detection.
  • High purity suitable for enzymatic and assay applications.
  • Available as powder and as 10 mM solution in DMSO.
  • Convenient for spectrophotometric quantification of enzyme activity.
Catalog No. 50-002-61771 Supplier Medchemexpress LLC Supplier No. HY13781510MG
4-Nitrophenyl α-L-rhamnopyranoside is a chromogenic substrate used to detect and quantify α-L-rhamnosidase activity. Enzymatic hydrolysis releases p-nitrophenol, producing a yellow product that is convenient for spectrophotometric measurement. Supplied in high purity as a powder, it is also available as a 10 mM solution in DMSO for ready use. For research use only.
  • Chromogenic substrate for α-L-rhamnosidase assays.
  • Releases p-nitrophenol on hydrolysis for colorimetric detection.
  • High purity suitable for enzymatic and assay applications.
  • Available as powder and as 10 mM solution in DMSO.
  • Convenient for spectrophotometric quantification of enzyme activity.