Learn More
Medchemexpress LLC 4-nitrophenyl-n-acetyl-α-d-galactosaminide | 23646-68-6 | MFCD00065476 | 99.9% | 342.30 g·mol⁻1 | C14H18N2O8 | 10 MG
SDP
Catalog No. 5000299674
Encompass_Preferred
4-Nitrophenyl-N-acetyl-α-D-galactosaminide (pNP-GalNAc) is a chromogenic substrate for N-acetyl-D-galactosaminase used in enzyme assays and kinetic studies. Enzymatic cleavage liberates 4-nitrophenol, producing a measurable absorbance change for quantitative activity determination.
  • Chromogenic substrate for N-acetyl-D-galactosaminase.
  • Enables quantitative measurement of enzyme activity by absorbance.
  • High reported purity (≈99.9%).
  • Solid form, white to off-white, suitable for standard assay preparation.
  • Store at 4°C and protect from light for optimal stability.
Catalog No. 50-002-99674 Supplier Medchemexpress LLC Supplier No. HYW01599610MG
4-Nitrophenyl-N-acetyl-α-D-galactosaminide (pNP-GalNAc) is a chromogenic substrate for N-acetyl-D-galactosaminase used in enzyme assays and kinetic studies. Enzymatic cleavage liberates 4-nitrophenol, producing a measurable absorbance change for quantitative activity determination.
  • Chromogenic substrate for N-acetyl-D-galactosaminase.
  • Enables quantitative measurement of enzyme activity by absorbance.
  • High reported purity (≈99.9%).
  • Solid form, white to off-white, suitable for standard assay preparation.
  • Store at 4°C and protect from light for optimal stability.